Assay Details
Gene Symbol: MYB85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118394.3 | NP_567664.1 | 1 - 2 | 409 | 165 | |
GenBank mRNA | AF175993.2 | - | 1 - 2 | 271 | 165 | |
AK175718.1 | - | 1 - 2 | 392 | 165 | ||
AK175777.1 | - | 1 - 2 | 392 | 165 | ||
AY085497.1 | - | 1 - 2 | 395 | 165 | ||
AY519608.1 | - | 1 - 2 | 259 | 165 | ||
DQ446863.1 | - | 1 - 2 | 259 | 165 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
