Assay Details
| Gene Symbol: PYR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341566.1 | NP_001327985.1 | 5 - 6 | 673 | 78 | |
| NM_118422.4 | NP_194024.1 | 5 - 6 | 673 | 78 | ||
| GenBank mRNA | AF000146.1 | - | 5 - 6 | 583 | 78 | |
More Information
Set Membership: |
|

PYR4
Genomic Map