Assay Details
| Gene Symbol: AGL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341568.1 | NP_001320037.1 | 6 - 7 | 811 | 110 | |
| NM_118424.2 | NP_194026.1 | 5 - 6 | 461 | 110 | ||
| GenBank mRNA | AF312664.1 | - | 5 - 6 | 461 | 110 | |
| AY142523.1 | - | 5 - 6 | 461 | 110 | ||
More Information
Set Membership: |
|

AGL19
Genomic Map