Assay Details
| Gene Symbol: CRK11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341581.1 | NP_001329689.1 | 2 - 3 | 1102 | 106 | |
| NM_001341582.1 | NP_001329690.1 | 3 - 4 | 993 | 106 | ||
| NM_001341583.1 | NP_001329691.1 | 2 - 3 | 745 | 106 | ||
| NM_118448.4 | NP_194050.2 | 2 - 3 | 1102 | 106 | ||
More Information
Set Membership: |
|

CRK11
Genomic Map