Assay Details
| Gene Symbol: CRK11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341582.1 | NP_001329690.1 | 7 - 8 | 1721 | 89 | |
| NM_001341583.1 | NP_001329691.1 | 6 - 7 | 1473 | 89 | ||
| NM_118448.4 | NP_194050.2 | 6 - 7 | 1830 | 89 | ||
More Information
Set Membership: |
|

CRK11
Genomic Map