Assay Details
| Gene Symbol: CRK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084966.2 | NP_001078435.1 | 4 - 5 | 1552 | 88 | |
| NM_001341586.1 | NP_001329248.1 | 4 - 5 | 1552 | 88 | ||
| NM_001341587.1 | NP_001329247.1 | 4 - 5 | 1552 | 88 | ||
| NM_118450.4 | NP_194052.2 | 4 - 5 | 1552 | 88 | ||
| NM_179096.3 | NP_849427.1 | 4 - 5 | 1552 | 88 | ||
| GenBank mRNA | AY142503.1 | - | 4 - 5 | 1332 | 88 | |
| BT001110.1 | - | 4 - 5 | 1332 | 88 | ||
| DQ680080.1 | - | 4 - 5 | 1393 | 88 | ||
More Information
Set Membership: |
|

CRK13
Genomic Map