Assay Details
| Gene Symbol: CRK22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118459.5 | NP_194061.2 | 1 - 2 | 1231 | 79 | |
| GenBank mRNA | AK176570.1 | - | 1 - 2 | 924 | 79 | |
| BT010572.1 | - | 1 - 2 | 854 | 79 | ||
More Information
Set Membership: |
|

CRK22
Genomic Map