Assay Details
| Gene Symbol: CRK22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118459.5 | NP_194061.2 | 6 - 7 | 2095 | 63 | |
| GenBank mRNA | AK176570.1 | - | 6 - 7 | 1788 | 63 | |
| BT010572.1 | - | 6 - 7 | 1718 | 63 | ||
More Information
Set Membership: |
|

CRK22
Genomic Map