Assay Details
| Gene Symbol: SGT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036627.2 | NP_001031704.1 | 7 - 8 | 896 | 63 | |
| NM_118488.6 | NP_194088.1 | 8 - 9 | 938 | 63 | ||
| NM_179098.3 | NP_849429.1 | 7 - 8 | 1022 | 63 | ||
| GenBank mRNA | AY063978.1 | - | 7 - 8 | 893 | 63 | |
| AY096395.1 | - | 7 - 8 | 756 | 63 | ||
More Information
Set Membership: |
|

SGT1A
Genomic Map