Assay Details
| Gene Symbol: EMB2816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126432.3 | NP_178480.1 | 2 - 3 | 181 | 92 | |
| NM_179600.3 | NP_849931.1 | 3 - 4 | 293 | 92 | ||
| GenBank mRNA | AK227358.1 | - | 3 - 4 | 267 | 92 | |
| BT003087.1 | - | 1 - 2 | 92 | 92 | ||
More Information
Set Membership: |
|

EMB2816
Genomic Map