Assay Details
| Gene Symbol: RTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341042.1 | NP_001328413.1 | 1 - 2 | 495 | 99 | |
| NM_148331.6 | NP_680697.4 | 1 - 2 | 456 | 99 | ||
More Information
Set Membership: |
|

RTL1
Genomic Map