Assay Details
| Gene Symbol: EMB1687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148433.3 | NP_683274.1 | 1 | 461 | 116 | |
| GenBank mRNA | BT015147.1 | - | 1 | 431 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMB1687| Gene Symbol: EMB1687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148433.3 | NP_683274.1 | 1 | 461 | 116 | |
| GenBank mRNA | BT015147.1 | - | 1 | 431 | 116 | |
Set Membership: |
|