Assay Details
| Gene Symbol: AT1G16225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332213.1 | NP_001323129.1 | 5 - 6 | 729 | 64 | |
| NM_001332214.1 | NP_001323130.1 | 4 - 5 | 854 | 64 | ||
| NM_148470.3 | NP_683311.2 | 4 - 5 | 551 | 64 | ||
More Information
Set Membership: |
|

AT1G16225
Genomic Map