Assay Details
| Gene Symbol: JAZ8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102753.4 | NP_564349.1 | 1 - 2 | 250 | 114 | |
| GenBank mRNA | AY087122.1 | - | 1 - 2 | 111 | 114 | |
| BT026098.1 | - | 1 - 2 | 86 | 114 | ||
More Information
Set Membership: |
|

JAZ8
Genomic Map