Assay Details
Gene Symbol: CIPK23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332883.1 | NP_001320726.1 | 11 - 12 | 1603 | 80 | |
NM_001332884.1 | NP_001320727.1 | 11 - 12 | 1739 | 80 | ||
NM_102766.6 | NP_564353.1 | 11 - 12 | 1657 | 80 | ||
GenBank mRNA | AY035226.1 | - | 11 - 12 | 1187 | 80 | |
AY056419.1 | - | 11 - 12 | 1559 | 80 | ||
AY090322.1 | - | 11 - 12 | 1187 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
