Assay Details
Gene Symbol: CIPK23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332883.1 | NP_001320726.1 | 1 - 2 | 643 | 140 | |
NM_001332884.1 | NP_001320727.1 | 1 - 2 | 640 | 140 | ||
NM_102766.6 | NP_564353.1 | 1 - 2 | 697 | 140 | ||
GenBank mRNA | AY035226.1 | - | 1 - 2 | 227 | 140 | |
AY056419.1 | - | 1 - 2 | 599 | 140 | ||
AY090322.1 | - | 1 - 2 | 227 | 140 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001332884.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
