Assay Details
| Gene Symbol: CIPK23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332883.1 | NP_001320726.1 | 5 - 6 | 963 | 73 | |
| NM_001332884.1 | NP_001320727.1 | 5 - 6 | 1099 | 73 | ||
| NM_102766.6 | NP_564353.1 | 5 - 6 | 1017 | 73 | ||
| GenBank mRNA | AY035226.1 | - | 5 - 6 | 547 | 73 | |
| AY056419.1 | - | 5 - 6 | 919 | 73 | ||
| AY090322.1 | - | 5 - 6 | 547 | 73 | ||
More Information
Set Membership: |
|

CIPK23
Genomic Map