Assay Details
| Gene Symbol: CIPK23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332883.1 | NP_001320726.1 | 8 - 9 | 1236 | 159 | |
| NM_001332884.1 | NP_001320727.1 | 8 - 9 | 1372 | 159 | ||
| NM_102766.6 | NP_564353.1 | 8 - 9 | 1290 | 159 | ||
| GenBank mRNA | AY035226.1 | - | 8 - 9 | 820 | 159 | |
| AY056419.1 | - | 8 - 9 | 1192 | 159 | ||
| AY090322.1 | - | 8 - 9 | 820 | 159 | ||
More Information
Set Membership: |
|

CIPK23
Genomic Map