Assay Details
| Gene Symbol: CIPK23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332883.1 | NP_001320726.1 | 9 - 10 | 1353 | 67 | |
| NM_001332884.1 | NP_001320727.1 | 9 - 10 | 1489 | 67 | ||
| NM_102766.6 | NP_564353.1 | 9 - 10 | 1407 | 67 | ||
| GenBank mRNA | AY035226.1 | - | 9 - 10 | 937 | 67 | |
| AY056419.1 | - | 9 - 10 | 1309 | 67 | ||
| AY090322.1 | - | 9 - 10 | 937 | 67 | ||
More Information
Set Membership: |
|

CIPK23
Genomic Map