Assay Details
| Gene Symbol: ARF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036038.2 | NP_001031115.1 | 12 - 13 | 1873 | 105 | |
| NM_001332891.1 | NP_001320924.1 | 11 - 12 | 2109 | 105 | ||
| NM_102771.4 | NP_174323.1 | 11 - 12 | 1978 | 105 | ||
| GenBank mRNA | AF013467.1 | - | 11 - 12 | 1903 | 105 | |
| AY669788.1 | - | 11 - 12 | 1157 | 105 | ||
More Information
Set Membership: |
|

ARF6
Genomic Map