Assay Details
| Gene Symbol: ARF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036038.2 | NP_001031115.1 | 5 - 6 | 1035 | 87 | |
| NM_001332891.1 | NP_001320924.1 | 4 - 5 | 1271 | 87 | ||
| NM_102771.4 | NP_174323.1 | 4 - 5 | 1140 | 87 | ||
| GenBank mRNA | AF013467.1 | - | 4 - 5 | 1065 | 87 | |
| AY669788.1 | - | 4 - 5 | 319 | 87 | ||
More Information
Set Membership: |
|

ARF6
Genomic Map