Assay Details
| Gene Symbol: ARF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036038.2 | NP_001031115.1 | 9 - 10 | 1528 | 97 | |
| NM_001332891.1 | NP_001320924.1 | 8 - 9 | 1764 | 97 | ||
| NM_102771.4 | NP_174323.1 | 8 - 9 | 1633 | 97 | ||
| GenBank mRNA | AF013467.1 | - | 8 - 9 | 1558 | 97 | |
| AY669788.1 | - | 8 - 9 | 812 | 97 | ||
More Information
Set Membership: |
|

ARF6
Genomic Map