Assay Details
| Gene Symbol: MUR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084163.1 | NP_001077632.1 | 2 - 3 | 449 | 109 | |
| NM_001332913.1 | NP_001319115.1 | 2 - 3 | 262 | 109 | ||
| NM_001332914.1 | NP_001321845.1 | 2 - 3 | 527 | 109 | ||
| NM_001332915.1 | NP_001321843.1 | 2 - 3 | 342 | 109 | ||
| NM_001332916.1 | NP_001321844.1 | 1 - 2 | 318 | 109 | ||
| NM_102799.6 | NP_174350.2 | 2 - 3 | 481 | 109 | ||
| GenBank mRNA | AF325108.1 | - | 1 - 2 | 58 | 109 | |
| AK317172.1 | - | 3 - 4 | 455 | 109 | ||
| AY064973.1 | - | 2 - 3 | 394 | 109 | ||
| AY195742.1 | - | 1 - 2 | 96 | 109 | ||
| BT002614.1 | - | 1 - 2 | 58 | 109 | ||
More Information
Set Membership: |
|

MUR4
Genomic Map