Assay Details
| Gene Symbol: MUR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084163.1 | NP_001077632.1 | 4 - 5 | 687 | 80 | |
| NM_001332913.1 | NP_001319115.1 | 4 - 5 | 503 | 80 | ||
| NM_001332914.1 | NP_001321845.1 | 4 - 5 | 768 | 80 | ||
| NM_001332915.1 | NP_001321843.1 | 4 - 5 | 583 | 80 | ||
| NM_001332916.1 | NP_001321844.1 | 3 - 4 | 559 | 80 | ||
| NM_102799.6 | NP_174350.2 | 4 - 5 | 722 | 80 | ||
| GenBank mRNA | AF325108.1 | - | 3 - 4 | 299 | 80 | |
| AK317172.1 | - | 5 - 6 | 696 | 80 | ||
| AY064973.1 | - | 4 - 5 | 635 | 80 | ||
| AY195742.1 | - | 3 - 4 | 337 | 80 | ||
| BT002614.1 | - | 3 - 4 | 299 | 80 | ||
More Information
Set Membership: |
|

MUR4
Genomic Map