Assay Details
| Gene Symbol: AGL86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102898.1 | NP_174444.1 | 1 | 742 | 133 | |
| GenBank mRNA | DQ056473.1 | - | 1 | 742 | 133 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL86| Gene Symbol: AGL86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102898.1 | NP_174444.1 | 1 | 742 | 133 | |
| GenBank mRNA | DQ056473.1 | - | 1 | 742 | 133 | |
Set Membership: |
|