Assay Details
| Gene Symbol: ROPGEF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332971.1 | NP_001320846.1 | 2 - 3 | 876 | 65 | |
| NM_102900.5 | NP_174446.2 | 4 - 5 | 1077 | 65 | ||
| GenBank mRNA | AK222010.1 | - | 4 - 5 | 1024 | 65 | |
More Information
Set Membership: |
|

ROPGEF14
Genomic Map