Assay Details
| Gene Symbol: ROPGEF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332971.1 | NP_001320846.1 | 5 - 6 | 1579 | 84 | |
| NM_102900.5 | NP_174446.2 | 7 - 8 | 1780 | 84 | ||
| GenBank mRNA | AK222010.1 | - | 7 - 8 | 1727 | 84 | |
More Information
Set Membership: |
|

ROPGEF14
Genomic Map