Assay Details
Gene Symbol: AT1G31760 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102910.4 | NP_564382.1 | 1 - 2 | 1099 | 78 | |
GenBank mRNA | AY085089.1 | - | 1 - 2 | 281 | 78 | |
BT024754.1 | - | 1 - 2 | 212 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
