Assay Details
| Gene Symbol: STRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102933.3 | NP_174479.1 | 11 - 12 | 1587 | 73 | |
| GenBank mRNA | AY080680.1 | - | 11 - 12 | 1483 | 73 | |
| AY117298.1 | - | 11 - 12 | 1429 | 73 | ||
More Information
Set Membership: |
|

STRS1
Genomic Map