Assay Details
| Gene Symbol: ATS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102953.4 | NP_174499.1 | 4 - 5 | 774 | 89 | |
| NM_179407.3 | NP_849738.1 | 4 - 5 | 774 | 89 | ||
| GenBank mRNA | AY093169.1 | - | 4 - 5 | 659 | 89 | |
| BT008758.1 | - | 4 - 5 | 601 | 89 | ||
More Information
Set Membership: |
|

ATS1
Genomic Map