Assay Details
| Gene Symbol: AOX1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102968.3 | NP_564395.1 | 3 - 4 | 1013 | 107 | |
| GenBank mRNA | BT029014.1 | - | 3 - 4 | 892 | 107 | |
More Information
Set Membership: |
|

AOX1D
Genomic Map