Assay Details
Gene Symbol: PRS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102972.3 | NP_174516.1 | 1 - 2 | 438 | 65 | |
GenBank mRNA | AF439851.1 | - | 1 - 2 | 420 | 65 | |
AY125555.1 | - | 1 - 2 | 139 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
