Assay Details
| Gene Symbol: ROPGEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120005.4 | NP_195556.1 | 4 - 5 | 1524 | 87 | |
| GenBank mRNA | AY056375.1 | - | 4 - 5 | 1345 | 87 | |
| AY084995.1 | - | 4 - 5 | 1441 | 87 | ||
| AY133649.1 | - | 4 - 5 | 1265 | 87 | ||
| BT000462.1 | - | 4 - 5 | 1345 | 87 | ||
| BT000975.1 | - | 4 - 5 | 1265 | 87 | ||
More Information
Set Membership: |
|

ROPGEF1
Genomic Map