Assay Details
| Gene Symbol: PLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120014.5 | NP_195565.2 | 3 - 4 | 777 | 85 | |
| GenBank mRNA | AK222239.1 | - | 3 - 4 | 670 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLC1| Gene Symbol: PLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120014.5 | NP_195565.2 | 3 - 4 | 777 | 85 | |
| GenBank mRNA | AK222239.1 | - | 3 - 4 | 670 | 85 | |
Set Membership: |
|