Assay Details
| Gene Symbol: PLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120014.5 | NP_195565.2 | 5 - 6 | 1286 | 78 | |
| GenBank mRNA | AK222239.1 | - | 5 - 6 | 1179 | 78 | |
More Information
Set Membership: |
|

PLC1
Genomic Map