Assay Details
Gene Symbol: BGAL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160820.1 | NP_001154292.1 | 11 - 12 | 1565 | 103 | |
NM_120020.4 | NP_195571.3 | 11 - 12 | 1629 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
