Assay Details
Gene Symbol: BGAL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160820.1 | NP_001154292.1 | 2 - 3 | 297 | 114 | |
NM_120020.4 | NP_195571.3 | 2 - 3 | 349 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
