Assay Details
| Gene Symbol: RPN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120024.3 | NP_195575.1 | 6 - 7 | 976 | 85 | |
| GenBank mRNA | AF360319.1 | - | 6 - 7 | 918 | 85 | |
| AY113889.1 | - | 5 - 6 | 837 | 85 | ||
| U33269.1 | - | 6 - 7 | 857 | 85 | ||
More Information
Set Membership: |
|

RPN10
Genomic Map