Assay Details
Gene Symbol: GRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120029.3 | NP_195580.1 | 1 | 1152 | 108 | |
GenBank mRNA | S47408.1 | - | 1 | 876 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
