Assay Details
Gene Symbol: VHA-A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120068.5 | NP_568051.1 | 10 - 11 | 1327 | 97 | |
GenBank mRNA | AK227321.1 | - | 10 - 11 | 1294 | 97 | |
AY060557.1 | - | 10 - 11 | 1288 | 97 | ||
BT002615.1 | - | 10 - 11 | 1213 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
