Assay Details
Gene Symbol: RD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120069.5 | NP_568052.1 | 1 - 2 | 750 | 73 | |
GenBank mRNA | AK226399.1 | - | 1 - 2 | 474 | 73 | |
AY080598.1 | - | 1 - 2 | 469 | 73 | ||
AY133844.1 | - | 1 - 2 | 460 | 73 |
Gene Symbol: AT2G24240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK229200.1 | - | 1838 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
