Assay Details
Gene Symbol: IMPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342523.1 | NP_001320164.1 | 2 - 3 | 427 | 86 | |
NM_120072.5 | NP_195623.3 | 2 - 3 | 514 | 86 | ||
GenBank mRNA | AK175533.1 | - | 2 - 3 | 455 | 86 | |
AK220687.1 | - | 2 - 3 | 455 | 86 | ||
BT010891.1 | - | 2 - 3 | 427 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
