Assay Details
Gene Symbol: NST-K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204038.1 | NP_001190967.1 | 6 - 7 | 1028 | 71 | |
NM_120099.4 | NP_568059.1 | 5 - 6 | 1015 | 71 | ||
NM_179196.2 | NP_849527.1 | 5 - 6 | 993 | 71 | ||
GenBank mRNA | AY091057.1 | - | 5 - 6 | 907 | 71 | |
AY142535.1 | - | 4 - 5 | 684 | 71 | ||
KJ667160.1 | - | 4 - 5 | 684 | 71 | ||
KP872771.1 | - | 4 - 5 | 684 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
