Assay Details
| Gene Symbol: NPY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335440.1 | NP_001318225.1 | 3 - 4 | 283 | 70 | |
| NM_127046.2 | NP_179089.1 | 2 - 3 | 722 | 70 | ||
More Information
Set Membership: |
|

NPY2
Genomic Map