Assay Details
| Gene Symbol: NPY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335440.1 | NP_001318225.1 | 4 - 5 | 1469 | 102 | |
| NM_127046.2 | NP_179089.1 | 3 - 4 | 1908 | 102 | ||
More Information
Set Membership: |
|

NPY2
Genomic Map