Assay Details
Gene Symbol: LIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127084.3 | NP_179126.2 | 1 - 2 | 324 | 98 | |
GenBank mRNA | AF530477.1 | - | 1 - 2 | 165 | 98 | |
AK229578.1 | - | 1 - 2 | 223 | 98 | ||
BT025547.1 | - | 1 - 2 | 138 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
