Assay Details
| Gene Symbol: FUT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127097.2 | NP_179139.1 | 1 - 2 | 209 | 143 | |
| GenBank mRNA | KY906035.1 | - | 1 - 2 | 156 | 143 | |
More Information
Set Membership: |
|

FUT5
Genomic Map