Assay Details
| Gene Symbol: HB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127165.4 | NP_179204.1 | 2 - 3 | 487 | 69 | |
| GenBank mRNA | AK227823.1 | - | 2 - 3 | 289 | 69 | |
| BT006405.1 | - | 2 - 3 | 232 | 69 | ||
More Information
Set Membership: |
|

HB1
Genomic Map