Assay Details
| Gene Symbol: PRORP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335484.1 | NP_001324533.1 | 4 - 5 | 1286 | 129 | |
| NM_127217.4 | NP_179256.3 | 3 - 4 | 1411 | 129 | ||
| GenBank mRNA | AK175948.1 | - | 3 - 4 | 1248 | 129 | |
More Information
Set Membership: |
|

PRORP2
Genomic Map