Assay Details
| Gene Symbol: PRORP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335484.1 | NP_001324533.1 | 5 - 6 | 1474 | 61 | |
| NM_127217.4 | NP_179256.3 | 4 - 5 | 1599 | 61 | ||
| GenBank mRNA | AK175948.1 | - | 4 - 5 | 1436 | 61 | |
More Information
Set Membership: |
|

PRORP2
Genomic Map